Ethyl-2,2-dimethyl-3-(2-methyl-1-propenyl)cyclopropane-1-carboxylate ≥95.0%

Supplier: TCI America

D2112-25G D2112-100G
TCD2112-025GEA 86.9 CAD
TCD2112-025G TCD2112-100G
Ethyl-2,2-dimethyl-3-(2-methyl-1-propenyl)cyclopropane-1-carboxylate ≥95.0%
Ethyl-2,2-dimethyl-3-(2-methyl-1-propenyl)cyclopropane-1-carboxylate

CAS Number: 97-41-6
MDL Number: MFCD00001304
Molecular Formula: C12H20O2
Molecular Weight: 196.29
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 112
Flash Point (°C): 84
Specific Gravity (20/20): 0.92


Product Categories: Bioscience, Monocyclic Monoterpenes
Other Product Names / Synonyms: Chrysanthemylmonocarboxylic Acid Ethyl Ester, Ethyl Chrysanthemate, Ethyl Chrysanthemumate, 2,2-Dimethyl-3-(2-methyl-1-propenyl)cyclopropane-1-carboxylic Acid Ethyl Ester

Formula: C₁₂H₂₀O₂
Boiling Pt: 112 °C
Density: 0.92 (20 °C)
Flash Pt: 84 °C
MDL Number: MFCD00001304
CAS Number: 97-41-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ccf313a70258856e8f8cd9b337d6c35e50a1b6fb&vid=ccf313a70258856e8f8cd9b337d6c35e50a1b6fb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR