2-Hydroxyethyl acetate 60.0% (by GC)
Supplier: TCI America
|
|
A1059-25ML
A1059-500ML
TCA1059-025MLEA
136.62
CAD
TCA1059-025ML
TCA1059-500ML
2-Hydroxyethyl acetate 60.0% (by GC)
2-Hydroxyethyl acetate
CAS Number: 542-59-6
MDL Number: MFCD00002864
Molecular Formula: C4H8O3
Molecular Weight: 104.11
Purity/Analysis Method: >60.0% (GC)
Form: Clear Liquid
Boiling point (°C): 182
Flash Point (°C): 102
Specific Gravity (20/20): 1.11
Other Product Names / Synonyms: Ethylene Glycol Monoacetate, Acetic Acid 2-Hydroxyethyl Ester
Formula:
C₄H₈O₃ Boiling Pt: 182 °C Density: 1.11 (20 °C) Flash Pt: 102 °C |
MDL Number:
MFCD00002864 CAS Number: 542-59-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...