2,2-Dimethyl-5-phenyl-1,3-dioxane-4,6-dione (5-phenylmeldrum's acid) ≥98.0%

Supplier: TCI America

Synonyms: 2,2-Dimethyl-5-phenyl-1,3-dioxane-4,6-dione, 5-Phenylmeldrum's acid

D2095-25G D2095-5G
TCD2095-025GEA 845.3 CAD
TCD2095-025G TCD2095-5G
2,2-Dimethyl-5-phenyl-1,3-dioxane-4,6-dione (5-phenylmeldrum's acid) ≥98.0%
2,2-Dimethyl-5-phenyl-1,3-dioxane-4,6-dione (5-Phenylmeldrum's acid)

CAS Number: 15231-78-4
MDL Number: MFCD00006636
Molecular Formula: C12H12O4
Molecular Weight: 220.22
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Other Product Names / Synonyms: 5-Phenylmeldrum's Acid

Formula: C₁₂H₁₂O₄
MDL Number: MFCD00006636
CAS Number: 15231-78-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d6315eda1962d5de939a73d29835d0f7db0c2d4&vid=8d6315eda1962d5de939a73d29835d0f7db0c2d4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR