Bis(2-ethylhexyl)dodecanedioate ≥94.0%

Supplier: TCI America

Synonyms: Dioctyl Dodecanedioate, Dodecanedioic Acid Di(2-ethylhexyl) Ester, Di(2-ethylhexyl) Dodecanedioate, Bis(2-ethylhexyl) dodecanedioate, Dodecanedioic Acid Dioctyl Ester

D2092-25ML D2092-500ML
TCD2092-025MLEA 45.82 CAD
TCD2092-025ML TCD2092-500ML
Bis(2-ethylhexyl)dodecanedioate ≥94.0%
Bis(2-ethylhexyl)dodecanedioate

CAS Number: 19074-24-9
MDL Number: MFCD00144216
Molecular Formula: C28H54O4
Molecular Weight: 454.74
Purity/Analysis Method: >94.0% (GC)
Form: Clear Liquid
Boiling point (°C): 267
Flash Point (°C): 230
Specific Gravity (20/20): 0.91


Other Product Names / Synonyms: Dodecanedioic Acid Di(2-ethylhexyl) Ester, Di(2-ethylhexyl) Dodecanedioate, Dioctyl Dodecanedioate, Dodecanedioic Acid Dioctyl Ester

Formula: C₂₈H₅₄O₄
MW: 454.74 g/mol
Boiling Pt: 267 °C
Density: 0.91 (20 °C)
Flash Pt: 230 °C
MDL Number: MFCD00144216
CAS Number: 19074-24-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d62bc85e87d6fc5398246ebfb7a38785dd5ab9d4&vid=d62bc85e87d6fc5398246ebfb7a38785dd5ab9d4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 94.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR