1,3-Diiminoisoindoline ≥98.0% (by titrimetric analysis)
Supplier: TCI America
D2079-25G
D2079-5G
TCD2079-025GEA
270.18
CAD
TCD2079-025G
TCD2079-5G
1,3-Diiminoisoindoline ≥98.0% (by titrimetric analysis)
1,3-Diiminoisoindoline
CAS Number: 57500-34-2
MDL Number: MFCD00010062
Molecular Formula: C8H7N3
Molecular Weight: 145.17
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 195
Formula:
C₈H₇N₃ Melting Pt: 195 °C |
MDL Number:
MFCD00010062 CAS Number: 57500-34-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...