3-Aminopyrrolidine dihydrochloride ≥98.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: Pyrrolidin-3-amine dihydrochloride

A1055-5G A1055-25G
TCA1055-005GEA 112.34 CAD
TCA1055-005G TCA1055-025G
3-Aminopyrrolidine dihydrochloride ≥98.0% (by total nitrogen basis)
3-Aminopyrrolidine dihydrochloride

CAS Number: 103831-11-4
MDL Number: MFCD00060176
Molecular Formula: C4H10N2
Molecular Weight: 159.05
Purity/Analysis Method: >98.0% (N)
Form: Crystal

Formula: C₄H₁₀N₂
MW: 159.05 g/mol
MDL Number: MFCD00060176
CAS Number: 103831-11-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e78b16c7236bcbfcc5534704914260815b226744&vid=e78b16c7236bcbfcc5534704914260815b226744&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR