1,4-Di(2-thienyl)-1,3-butadiene (mixture of isomers) ≥97.0%

Supplier: TCI America

D2077-1G D2077-5G
TCD2077-001GEA 262.75 CAD
TCD2077-001G TCD2077-005G
1,4-Di(2-thienyl)-1,3-butadiene (mixture of isomers) ≥97.0%
1,4-Di(2-thienyl)-1,3-butadiene (mixture of isomers)

CAS Number: 23354-93-0
MDL Number: MFCD00142549
Molecular Formula: C12H10S2
Molecular Weight: 218.33
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 171

Formula: C₁₂H₁₀S₂
Melting Pt: 171 °C
MDL Number: MFCD00142549
CAS Number: 23354-93-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=33b322856b64592eb011b678fbf9b45552f22473&vid=33b322856b64592eb011b678fbf9b45552f22473&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR