1,11-Diaminoundecane ≥98.0%

Supplier: TCI America

Synonyms: 1,11-Undecanediamine, Undecamethylenediamine

D2075-5G D2075-1G
TCD2075-005GEA 344.44 CAD
TCD2075-005G TCD2075-001G
1,11-Diaminoundecane ≥98.0%
1,11-Diaminoundecane

CAS Number: 822-08-2
MDL Number: MFCD00142358
Molecular Formula: C11H26N2
Molecular Weight: 186.34
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 152
Melting point (°C): 45
Storage Temperature: 0-10°C


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Undecamethylenediamine, 1,11-Undecanediamine

Formula: C₁₁H₂₆N₂
MW: 186.34 g/mol
Boiling Pt: 152 °C
Melting Pt: 45 °C
MDL Number: MFCD00142358
CAS Number: 822-08-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3c3110e92bb4ae5f16b3a5077b98ea87a79b9178&vid=3c3110e92bb4ae5f16b3a5077b98ea87a79b9178&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR