trans-1,2-Di(2-thienyl)ethylene ≥98.0%
Supplier: TCI America
D2074-1G
D2074-5G
TCD2074-001GEA
206.66
CAD
TCD2074-001G
TCD2074-005G
trans-1,2-Di(2-thienyl)ethylene ≥98.0%
trans-1,2-Di(2-thienyl)ethylene
CAS Number: 13640-78-3
MDL Number: MFCD00145416
Molecular Formula: C10H8S2
Molecular Weight: 192.29
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 133
Formula:
C₁₀H₈S₂ Melting Pt: 133 °C |
MDL Number:
MFCD00145416 CAS Number: 13640-78-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...