Methyl-3-(dimethylamino)benzoate ≥99.0% (by titrimetric analysis)

Supplier: TCI America

D2069-10G D2069-25G
TCD2069-010GEA 140.62 CAD
TCD2069-010G TCD2069-25G
Methyl-3-(dimethylamino)benzoate ≥99.0% (by titrimetric analysis)
Methyl 3-(dimethylamino)benzoate

CAS Number: 16518-64-2
MDL Number: MFCD00144731
Molecular Formula: C10H13NO2
Molecular Weight: 179.22
Purity/Analysis Method: >99.0% (T)
Form: Clear Liquid
Boiling point (°C): 80
Specific Gravity (20/20): 1.10


Other Product Names / Synonyms: 3-(Dimethylamino)benzoic Acid Methyl Ester

Formula: C₁₀H₁₃NO₂
Boiling Pt: 80 °C
Density: 1.10 (20 °C)
MDL Number: MFCD00144731
CAS Number: 16518-64-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c015b36108642c2c2a9f12b8712e86c24776eb5&vid=0c015b36108642c2c2a9f12b8712e86c24776eb5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR