[2-(1,3-Dioxolan-2-yl)ethyl]triphenylphosphonium bromide ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: (2-(1,3-Dioxolan-2-yl)ethyl)triphenylphosphonium bromide

D2056-5G D2056-25G
TCD2056-5GEA 167.48 CAD
TCD2056-5G TCD2056-025G
[2-(1,3-Dioxolan-2-yl)ethyl]triphenylphosphonium bromide ≥97.0% (by titrimetric analysis)
[2-(1,3-Dioxolan-2-yl)ethyl]triphenylphosphonium bromide

CAS Number: 86608-70-0
MDL Number: MFCD00075119
Molecular Formula: C23H24BrO2P
Molecular Weight: 443.32
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 155

Formula: C₂₃H₂₄BrO₂P
MW: 443.3132 g/mol
Melting Pt: 155 °C
MDL Number: MFCD00075119
CAS Number: 86608-70-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=17eb81e4e93d6d5982675b4f43898b15ff7c1271&vid=17eb81e4e93d6d5982675b4f43898b15ff7c1271&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR