4,4'-Diisocyanato-3,3'-dimethylbiphenyl ≥98.0%

Supplier: TCI America

Synonyms: 3,3'-Dimethylbiphenyl-4,4'-diyl Diisocyanate

D2051-250G D2051-25G
TCD2051-250GEA 687.3 CAD
TCD2051-250G TCD2051-25G
4,4'-Diisocyanato-3,3'-dimethylbiphenyl ≥98.0%
4,4'-Diisocyanato-3,3'-dimethylbiphenyl

CAS Number: 91-97-4
MDL Number: MFCD00019908
Molecular Formula: C16H12N2O2
Molecular Weight: 264.28
Purity/Analysis Method: >98.0% (GC)
Form: Pellet
Melting point (°C): 72
Flash Point (°C): 218
Storage Temperature: 0-10°C


Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials, Biphenyls (for High-Performance Polymer Research)
Other Product Names / Synonyms: 3,3'-Dimethylbiphenyl-4,4'-diyl Diisocyanate

Formula: C₁₆H₁₂N₂O₂
Melting Pt: 72 °C
Flash Pt: 218 °C
MDL Number: MFCD00019908
CAS Number: 91-97-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d105143e7166ea1e08223a89b35f070a0bf1fcba&vid=d105143e7166ea1e08223a89b35f070a0bf1fcba&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR