trans-N-(6,6-Dimethyl-2-hepten-4-ynyl)-N-methyl-1-naphthylmethylamine hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
Terbinafine Hydrochloride, Terbinafine HCl, (E)-N,6,6-Trimethyl-N-(naphthalen-1-ylmethyl)hept-2-en-4-yn-1-amine hydrochloride
D2049-5G
D2049-1G
TCD2049-5GEA
446.35
CAD
TCD2049-5G
TCD2049-001G
trans-N-(6,6-Dimethyl-2-hepten-4-ynyl)-N-methyl-1-naphthylmethylamine hydrochloride ≥98.0% (by HPLC, titration analysis)
trans-N-(6,6-Dimethyl-2-hepten-4-ynyl)-N-methyl-1-naphthylmethylamine hydrochloride
CAS Number: 78628-80-5
MDL Number: MFCD00145430
Molecular Formula: C21H25N
Molecular Weight: 327.90
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 214
Product Categories: Bioscience, Antifungal Ingredients
Other Product Names / Synonyms: Terbinafine Hydrochloride
Formula:
C₂₁H₂₅N MW: 327.90 g/mol Melting Pt: 214 °C |
MDL Number:
MFCD00145430 CAS Number: 78628-80-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...