Sodium Diphenylphosphinobenzene-3-sulfonate ≥90.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Diphenylphosphinobenzene-3-sulfonic acid sodium salt, Sodium 3-(diphenylphosphino)benzenesulphonate, Triphenylphosphine-3-sulfonic acid sodium salt, Sodium 3-(diphenylphosphino)benzenesulfonate, Diphenylphosphinobenzene-3-sulphonic acid sodium salt, Triphenylphosphine-3-sulphonic acid sodium salt

D2043-1G D2043-5G
TCD2043-001GEA 208.54 CAD
TCD2043-001G TCD2043-005G
Sodium Diphenylphosphinobenzene-3-sulfonate ≥90.0% (by titrimetric analysis)
Sodium Diphenylphosphinobenzene-3-sulfonate

CAS Number: 63995-75-5
MDL Number: MFCD00144157
Molecular Formula: C18H15O3PS
Molecular Weight: 364.33
Purity/Analysis Method: >90.0% (T)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Cross-coupling Reaction using Transition Metal Catalysts, Ligands
Other Product Names / Synonyms: Diphenylphosphinobenzene-3-sulfonic Acid Sodium Salt, Triphenylphosphine-3-sulfonic Acid Sodium Salt

Formula: C₁₈H₁₅O₃PS
MW: 364.33 g/mol
MDL Number: MFCD00144157
CAS Number: 63995-75-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7656cb50f30f08e1d10f33d35fc140d05e508f6d&vid=eda66e21c6cdaec06e89bb813be87705a9065cdd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR