3,4-Diacetoxybenzaldehyde ≥98.0%
Supplier: TCI America
D2042-5G
D2042-25G
TCD2042-5GEA
181.7
CAD
TCD2042-5G
TCD2042-025G
3,4-Diacetoxybenzaldehyde ≥98.0%
3,4-Diacetoxybenzaldehyde
CAS Number: 67727-64-4
MDL Number: MFCD00143057
Molecular Formula: C11H10O5
Molecular Weight: 222.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 53
Formula:
C₁₁H₁₀O₅ Melting Pt: 53 °C |
MDL Number:
MFCD00143057 CAS Number: 67727-64-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...