Dodecamethylcyclohexasiloxane ≥97.0%

Supplier: TCI America

D2040-5ML D2040-1ML D2040-25ML
TCD2040-005MLEA 629.66 CAD
TCD2040-005ML TCD2040-1ML TCD2040-25ML
Dodecamethylcyclohexasiloxane ≥97.0%
Dodecamethylcyclohexasiloxane

CAS Number: 540-97-6
MDL Number: MFCD00144215
Molecular Formula: C12H36O6Si6
Molecular Weight: 444.92
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 245
Flash Point (°C): 105
Specific Gravity (20/20): 0.97

Formula: C₁₂H₃₆O₆Si₆
Boiling Pt: 245 °C
Density: 0.97 (20 °C)
Flash Pt: 105 °C
MDL Number: MFCD00144215
CAS Number: 540-97-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7e1c1df697dfb2d8409fc6a33eda2495e82d20cd&vid=7e1c1df697dfb2d8409fc6a33eda2495e82d20cd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR