Diphenyl (2,3-dihydro-2-thioxo-3-benzoxazolyl)phosphonate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

D2038-25G D2038-5G
TCD2038-25GEA 568.8 CAD
TCD2038-25G TCD2038-005G
Diphenyl (2,3-dihydro-2-thioxo-3-benzoxazolyl)phosphonate ≥98.0% (by titrimetric analysis)
Diphenyl (2,3-dihydro-2-thioxo-3-benzoxazolyl)phosphonate

CAS Number: 111160-56-6
MDL Number: MFCD00142793
Molecular Formula: C19H14NO4PS
Molecular Weight: 383.36
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 84


Product Categories: Synthetic Organic Chemistry, Condensation & Active Esterification
Other Product Names / Synonyms: (2,3-Dihydro-2-thioxo-3-benzoxazolyl)phosphonic Acid Diphenyl Ester, DBOP

Formula: C₁₉H₁₄NO₄PS
Melting Pt: 84 °C
MDL Number: MFCD00142793
CAS Number: 111160-56-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0e9ea8065cec09c719ab3f9f440eec79d63d825c&vid=0e9ea8065cec09c719ab3f9f440eec79d63d825c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR