3,5-Di(2-pyridyl)pyrazole ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2,2'-(1H-Pyrazole-3,5-diyl)dipyridine

D2037-1G D2037-5G
TCD2037-001GEA 369.72 CAD
TCD2037-001G TCD2037-5G
3,5-Di(2-pyridyl)pyrazole ≥98.0% (by GC, titration analysis)
3,5-Di(2-pyridyl)pyrazole

CAS Number: 129485-83-2
MDL Number: MFCD00130125
Molecular Formula: C13H10N4
Molecular Weight: 222.25
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 188

Formula: C₁₃H₁₀N₄
MW: 222.25 g/mol
Melting Pt: 188 °C
MDL Number: MFCD00130125
CAS Number: 129485-83-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a368e511108c7f30047303bb311dd7342524347a&vid=a368e511108c7f30047303bb311dd7342524347a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR