2,2-Di(2-tetrahydrofuryl)propane ≥96.0%
Supplier: TCI America
D2034-25ML
D2034-500ML
D2034-100ML
TCD2034-025MLEA
67.94
CAD
TCD2034-025ML
TCD2034-500ML
TCD2034-100ML
2,2-Di(2-tetrahydrofuryl)propane ≥96.0%
2,2-Di(2-tetrahydrofuryl)propane
CAS Number: 89686-69-1
MDL Number: MFCD00142763
Molecular Formula: C11H20O2
Molecular Weight: 184.28
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 146
Flash Point (°C): 95
Specific Gravity (20/20): 1.00
Formula:
C₁₁H₂₀O₂ Boiling Pt: 146 °C Density: 1.00 (20 °C) Flash Pt: 95 °C |
MDL Number:
MFCD00142763 CAS Number: 89686-69-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...