1,3-Dimethyl-5-pyrazolone ≥98.0%

Supplier: TCI America

Synonyms: 2,4-Dihydro-2,5-dimethyl-3H-pyrazol-3-one, 1,3-Dimethyl-1H-pyrazol-5(4H)-one

D2031-25G D2031-500G D2031-100G
TCD2031-25GEA 91.64 CAD
TCD2031-25G TCD2031-500G TCD2031-100G
1,3-Dimethyl-5-pyrazolone ≥98.0%
1,3-Dimethyl-5-pyrazolone

CAS Number: 2749-59-9
MDL Number: MFCD00067008
Molecular Formula: C5H8N2O
Molecular Weight: 112.13
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 117


Other Product Names / Synonyms: 2,4-Dihydro-2,5-dimethyl-3H-pyrazol-3-one

Formula: C₅H₈N₂O
MW: 112.13 g/mol
Melting Pt: 117 °C
MDL Number: MFCD00067008
CAS Number: 2749-59-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13d294b47276c40ecc6242248bdfc0f38b061ee8&vid=13d294b47276c40ecc6242248bdfc0f38b061ee8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR