1,4-Diiodo-2,3,5,6-tetramethylbenzene ≥95.0%
Supplier: TCI America
D2028-5G
D2028-25G
TCD2028-5GEA
161.95
CAD
TCD2028-5G
TCD2028-025G
1,4-Diiodo-2,3,5,6-tetramethylbenzene ≥95.0%
1,4-Diiodo-2,3,5,6-tetramethylbenzene
CAS Number: 3268-21-1
MDL Number: MFCD00142554
Molecular Formula: C10H12I2
Molecular Weight: 386.01
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 140
Other Product Names / Synonyms: 3,6-Diiododurene
Formula:
C₁₀H₁₂I₂ Melting Pt: 140 °C |
MDL Number:
MFCD00142554 CAS Number: 3268-21-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...