2,5-Diamino-1,4-benzenedithiol dihydrochloride ≥97.0%

Supplier: TCI America

Synonyms: 2,5-Dimercapto-1,4-phenylenediamine Dihydrochloride, 2,5-Diaminobenzene-1,4-dithiol dihydrochloride

D2022-25G D2022-5G
TCD2022-25GEA 2682.84 CAD
TCD2022-25G TCD2022-005G
2,5-Diamino-1,4-benzenedithiol dihydrochloride ≥97.0%
2,5-Diamino-1,4-benzenedithiol dihydrochloride

CAS Number: 75464-52-7
MDL Number: MFCD00142835
Molecular Formula: C6H8N2S2
Molecular Weight: 245.18
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Melting point (°C): 210


Product Categories: Materials Chemistry, Ligands for Functional Metal Complexes, Monomers, Polyimide Raw Materials
Other Product Names / Synonyms: 2,5-Dimercapto-1,4-phenylenediamine Dihydrochloride

Formula: C₆H₈N₂S₂
MW: 245.18 g/mol
Melting Pt: 210 °C
MDL Number: MFCD00142835
CAS Number: 75464-52-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a8b840a1e3006ddc7d736ce6974d522db1dfa30d&vid=a8b840a1e3006ddc7d736ce6974d522db1dfa30d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR