1,6-Diaminohexane-N,N,N',N'-tetraacetic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Hexamethylenediaminetetraacetic Acid
D2019-25G
D2019-5G
TCD2019-25GEA
1023.84
CAD
TCD2019-25G
76218-992
1,6-Diaminohexane-N,N,N',N'-tetraacetic acid ≥98.0% (by titrimetric analysis)
1,6-Diaminohexane-N,N,N',N'-tetraacetic acid
CAS Number: 1633-00-7
MDL Number: MFCD00004292
Molecular Formula: C14H24N2O8
Molecular Weight: 348.35
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 236
Product Categories: Analytical Science, Complexones (Chelating Agents)
Other Product Names / Synonyms: Hexamethylenediaminetetraacetic Acid
Formula:
C₁₄H₂₄N₂O₈ MW: 348.35 g/mol Melting Pt: 236 °C |
MDL Number:
MFCD00004292 CAS Number: 1633-00-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...