Sodium-2,6-dinitro-4-(trifluoromethyl)benzenesulfonate ≥95.0% (by titrimetric analysis)

Supplier: TCI America

D2011-25G D2011-5G
TCD2011-25GEA 736.28 CAD
TCD2011-25G TCD2011-005G
Sodium-2,6-dinitro-4-(trifluoromethyl)benzenesulfonate ≥95.0% (by titrimetric analysis)
Sodium-2,6-dinitro-4-(trifluoromethyl)benzenesulfonate

CAS Number: 54495-25-9
MDL Number: MFCD00059068
Molecular Formula: C7H3F3N2O7S
Molecular Weight: 338.14
Purity/Analysis Method: >95.0% (T)
Form: Crystal


Other Product Names / Synonyms: 2,6-Dinitro-4-(trifluoromethyl)benzenesulfonic Acid Sodium Salt

Formula: C₇H₃F₃N₂O₇S
MDL Number: MFCD00059068
CAS Number: 54495-25-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9ca3584312dbcda280e5b1b4133eecf1500e8901&vid=9ca3584312dbcda280e5b1b4133eecf1500e8901&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR