Ethyl-3,3-diethoxyacrylate ≥95.0%

Supplier: TCI America

D2006-5G D2006-25G
TCD2006-005GEA 469.42 CAD
TCD2006-005G TCD2006-25G
Ethyl-3,3-diethoxyacrylate ≥95.0%
Ethyl-3,3-diethoxyacrylate

CAS Number: 32002-24-7
MDL Number: MFCD00012220
Molecular Formula: C9H16O4
Molecular Weight: 188.22
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 132
Flash Point (°C): 102
Specific Gravity (20/20): 1.03


Other Product Names / Synonyms: 3,3-Diethoxyacrylic Acid Ethyl Ester

Formula: C₉H₁₆O₄
Boiling Pt: 132 °C
Density: 1.03 (20 °C)
Flash Pt: 102 °C
MDL Number: MFCD00012220
CAS Number: 32002-24-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bb99f524b341cd86c9593b4b65ce06182ee752e8&vid=bb99f524b341cd86c9593b4b65ce06182ee752e8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR