S,S-Diphenylsulfilimine monohydrate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

D2002-1G D2002-5G
TCD2002-1GEA 274.92 CAD
TCD2002-1G TCD2002-005G
S,S-Diphenylsulfilimine monohydrate ≥98.0% (by titrimetric analysis)
S,S-Diphenylsulfilimine monohydrate

CAS Number: 68837-61-6
MDL Number: MFCD00001757
Molecular Formula: C12H11NS
Molecular Weight: 201.29
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 71
Flash Point (°C): 110

Formula: C₁₂H₁₁NS
Melting Pt: 71 °C
Flash Pt: 110 °C
MDL Number: MFCD00001757
CAS Number: 68837-61-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8f0ef1fa1c8d5bcf9cb465f895e92eabfcf99a09&vid=8f0ef1fa1c8d5bcf9cb465f895e92eabfcf99a09&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR