2-Aminophenol-4-sulfonamide ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 3-Amino-4-hydroxybenzenesulphonamide, 3-Amino-4-hydroxybenzenesulfonamide

A1031-500G A1031-25G
TCA1031-500GEA 507.18 CAD
TCA1031-500G TCA1031-025G
2-Aminophenol-4-sulfonamide ≥97.0% (by HPLC, titration analysis)
2-Aminophenol-4-sulphonamide

CAS Number: 98-32-8
MDL Number: MFCD00035782
Molecular Formula: C6H8N2O3S
Molecular Weight: 188.20
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Melting point (°C): 199


Other Product Names / Synonyms: 3-Amino-4-hydroxybenzenesulfonamide, 4-Hydroxymetanilamide

Formula: C₆H₈N₂O₃S
MW: 188.2 g/mol
Melting Pt: 199 °C
MDL Number: MFCD00035782
CAS Number: 98-32-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0534a6cc6d3fe1bf39a9f51effeceaaf5d819ae1&vid=0534a6cc6d3fe1bf39a9f51effeceaaf5d819ae1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR