4-(2',6'-Dimethylphenoxy)phthalonitrile ≥99.0%

Supplier: TCI America

D1983-1G D1983-5G
TCD1983-001GEA 181.7 CAD
TCD1983-001G TCD1983-005G
4-(2',6'-Dimethylphenoxy)phthalonitrile ≥99.0%
4-(2',6'-Dimethylphenoxy)phthalonitrile

CAS Number: 221302-75-6
MDL Number: MFCD00059064
Molecular Formula: C16H12N2O
Molecular Weight: 248.29
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 110


Product Categories: Materials Chemistry, Building Blocks for Phthalocyanines
Other Product Names / Synonyms: 1,2-Dicyano-4-(2',6'-dimethylphenoxy)benzene

Formula: C₁₆H₁₂N₂O
Melting Pt: 110 °C
MDL Number: MFCD00059064
CAS Number: 221302-75-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2df8ed6df42d4bf59e1efb5c95af22fd63d20675&vid=2df8ed6df42d4bf59e1efb5c95af22fd63d20675&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR