Dimethyldioctadecylammonium bromide (DDA) ≥98.0% (by titrimetric analysis)
Supplier: TCI America
D1975-250G
D1975-25G
TCD1975-250GEA
826.66
CAD
TCD1975-250G
TCD1975-025G
Dimethyldioctadecylammonium bromide (DDA) ≥98.0% (by titrimetric analysis)
Dimethyldioctadecylammonium bromide (DDA)
CAS Number: 3700-67-2
MDL Number: MFCD00041975
Molecular Formula: C38H80BrN
Molecular Weight: 630.97
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 155
Other Product Names / Synonyms: Dimethyldistearylammonium Bromide
Formula:
C₃₈H₈₀BrN Melting Pt: 155 °C |
MDL Number:
MFCD00041975 CAS Number: 3700-67-2 EINECS: 223-037-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...