2,4-Dimethyl-2,4-pentanediol ≥96.0%

Supplier: TCI America

D1946-5ML D1946-25ML
TCD1946-005MLEA 556.16 CAD
TCD1946-005ML TCD1946-025ML
2,4-Dimethyl-2,4-pentanediol ≥96.0%
2,4-Dimethyl-2,4-pentanediol

CAS Number: 24892-49-7
MDL Number: MFCD00012306
Molecular Formula: C7H16O2
Molecular Weight: 132.20
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 100
Flash Point (°C): 85
Freezing point (°C): 8
Specific Gravity (20/20): 0.92

Formula: C₇H₁₆O₂
MW: 132.20 g/mol
Boiling Pt: 100 °C
Density: 0.92 (20 °C)
Flash Pt: 85 °C
MDL Number: MFCD00012306
CAS Number: 24892-49-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1df9b5d46c1e4989c2d0a9c2b7fae2106fd20baa&vid=1df9b5d46c1e4989c2d0a9c2b7fae2106fd20baa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR