2,4'-Dihydroxydiphenylmethane ≥98.0%

Supplier: TCI America

D1940-1G D1940-5G
TCD1940-001GEA 261.02 CAD
TCD1940-001G TCD1940-005G
2,4'-Dihydroxydiphenylmethane ≥98.0%
2,4'-Dihydroxydiphenylmethane

CAS Number: 2467-03-0
MDL Number: MFCD00060113
Molecular Formula: C13H12O2
Molecular Weight: 200.24
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 119

Formula: C₁₃H₁₂O₂
Melting Pt: 119 °C
MDL Number: MFCD00060113
CAS Number: 2467-03-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fc90bc1faabe6526fc5a781ecf1f31efb28b1cef&vid=fc90bc1faabe6526fc5a781ecf1f31efb28b1cef&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR