2,2'-Dihydroxydiphenylmethane ≥99.0%

Supplier: TCI America

D1939-5G D1939-1G
TCD1939-005GEA 499.85 CAD
TCD1939-005G 76218-990
2,2'-Dihydroxydiphenylmethane ≥99.0%
2,2'-Dihydroxydiphenylmethane

CAS Number: 2467-02-9
MDL Number: MFCD00002243
Molecular Formula: C13H12O2
Molecular Weight: 200.24
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 118


Product Categories: Materials Chemistry, Monomers, Diphenylmethanes (for High-Performance Polymer Research)
Other Product Names / Synonyms: Bis(o-hydroxyphenyl)methane, 2,2'-Methylenediphenol

Formula: C₁₃H₁₂O₂
Melting Pt: 118 °C
MDL Number: MFCD00002243
CAS Number: 2467-02-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a8a0a482a138d8cd091cc2e6c2557b2f64149929&vid=a8a0a482a138d8cd091cc2e6c2557b2f64149929&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR