9-Decenoic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

D1932-25ML D1932-5ML
TCD1932-25MLEA 495.54 CAD
TCD1932-25ML TCD1932-005ML
9-Decenoic acid ≥98.0% (by titrimetric analysis)
9-Decenoic acid

CAS Number: 14436-32-9
MDL Number: MFCD00036663
Molecular Formula: C10H18O2
Molecular Weight: 170.25
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Boiling point (°C): 162
Flash Point (°C): 110
Specific Gravity (20/20): 0.92

Formula: C₁₀H₁₈O₂
MW: 170.25 g/mol
Boiling Pt: 162 °C
Density: 0.92 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00036663
CAS Number: 14436-32-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=addd69d443dbb8e2a44af073ba0ad5d3746aaac7&vid=addd69d443dbb8e2a44af073ba0ad5d3746aaac7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR