3,4'-Diaminodiphenyl ether ≥97.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3-(4-Aminophenoxy)aniline, 3,4'-Oxydianiline

D1924-5G D1924-25G
TCD1924-5GEA 105.86 CAD
TCD1924-5G TCD1924-025G
3,4'-Diaminodiphenyl ether ≥97.0% (by GC, titration analysis)
3,4'-Diaminodiphenyl ether

CAS Number: 2657-87-6
MDL Number: MFCD00036097
Molecular Formula: C12H12N2O
Molecular Weight: 200.24
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Boiling point (°C): 207


Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials, Diphenyl Ethers (for High-Performance Polymer Research)
Other Product Names / Synonyms: 3,4'-Oxydianiline

Formula: C₁₂H₁₂N₂O
MW: 200.2365 g/mol
Boiling Pt: 207 °C
MDL Number: MFCD00036097
CAS Number: 2657-87-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=545d746ebd9fd6094eb8e9ccc0d0002f3a4e8b56&vid=545d746ebd9fd6094eb8e9ccc0d0002f3a4e8b56&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR