5-(2,5-Dioxotetrahydrofuryl)-3-methyl-3-cyclohexene-1,2-dicarboxylic anhydride ≥95.0%

Supplier: TCI America

Synonyms: 6-(2,5-Dioxotetrahydrofuran-3-yl)-4-methyl-7,7a-dihydroisobenzofuran-1,3(3aH,6H)-dione

D1901-25G D1901-500G
TCD1901-025GEA 116.92 CAD
TCD1901-025G TCD1901-500G
5-(2,5-Dioxotetrahydrofuryl)-3-methyl-3-cyclohexene-1,2-dicarboxylic anhydride ≥95.0%
5-(2,5-Dioxotetrahydrofuryl)-3-methyl-3-cyclohexene-1,2-dicarboxylic anhydride

CAS Number: 73003-90-4
MDL Number: MFCD00039141
Molecular Formula: C13H12O6
Molecular Weight: 264.23
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 174

Formula: C₁₃H₁₂O₆
Melting Pt: 174 °C
MDL Number: MFCD00039141
CAS Number: 73003-90-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d1bfeb643a5881d9e2f833d9d1c0dd26e51987a&vid=4d1bfeb643a5881d9e2f833d9d1c0dd26e51987a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR