4,6-Diaminoresorcinol dihydrochloride ≥98.0% (by total nitrogen and titration analysis)

Supplier: TCI America

D1888-25G D1888-5G
TCD1888-25GEA 439.71 CAD
TCD1888-25G TCD1888-005G
4,6-Diaminoresorcinol dihydrochloride ≥98.0% (by total nitrogen and titration analysis)
4,6-Diaminoresorcinol dihydrochloride

CAS Number: 16523-31-2
MDL Number: MFCD00143239
Molecular Formula: C6H8N2O2
Molecular Weight: 213.06
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal


Product Categories: Materials Chemistry, Ligands for Functional Metal Complexes, Monomers, Polyimide Raw Materials
Other Product Names / Synonyms: 4,6-Dihydroxy-1,3-phenylenediamine Dihydrochloride

Formula: C₆H₈N₂O₂
MDL Number: MFCD00143239
CAS Number: 16523-31-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=00e86e5fbb418bb9ed346f70e5f368802991e22d&vid=00e86e5fbb418bb9ed346f70e5f368802991e22d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR