9,10-Dihydro-9-oxa-10-phosphaphenanthrene-10-oxide ≥97.0%

Supplier: TCI America

Synonyms: 3,4:5,6-Dibenzo-1,2-oxaphosphane-2-oxide, 6H-Dibenzo[c,e][1,2]oxaphosphinine 6-oxide

D1874-25G D1874-500G D1874-100G
TCD1874-025GEA 53.06 CAD
TCD1874-025G TCD1874-500G TCD1874-100G
9,10-Dihydro-9-oxa-10-phosphaphenanthrene-10-oxide ≥97.0%
9,10-Dihydro-9-oxa-10-phosphaphenanthrene-10-oxide

CAS Number: 35948-25-5
MDL Number: MFCD00040561
Molecular Formula: C12H9O2P
Molecular Weight: 216.18
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 119
Flash Point (°C): 222


Other Product Names / Synonyms: 3,4:5,6-Dibenzo-1,2-oxaphosphane-2-oxide

Formula: C₁₂H₉O₂P
MW: 216.18 g/mol
Melting Pt: 119 °C
Flash Pt: 222 °C
MDL Number: MFCD00040561
CAS Number: 35948-25-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8640ef491ac04f63f894b670355d729ac6088d3c&vid=8640ef491ac04f63f894b670355d729ac6088d3c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR