Pentasodium diethylenetriaminepentaacetate ∼40% ∼1.0 mol/L in water

Supplier: TCI America

D1870-25G D1870-500G
TCD1870-25GEA 30.02 CAD
TCD1870-25G TCD1870-500G
Pentasodium diethylenetriaminepentaacetate ∼40% ∼1.0 mol/L in water
Pentasodium diethylenetriaminepentaacetate

CAS Number: 140-01-2
MDL Number: MFCD00051016
Molecular Formula: C14H23N3O10
Molecular Weight: 503.26
Form: Clear Liquid
Color: Slightly Pale Yellow
Specific Gravity (20/20): 1.30


Other Product Names / Synonyms: Diethylenetriaminepentaacetic Acid Pentasodium Salt

Formula: C₁₄H₂₃N₃O₁₀
Density: 1.30 (20 °C)
MDL Number: MFCD00051016
CAS Number: 140-01-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=145eff48412750378428a6ac86ccc7a72253ea69&vid=06206f7accf858111ecc285338f356bf0b0187c3&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR