Diethylenetriaminepentaacetic acid calcium trisodium salt hydrate ≥98.0%

Supplier: TCI America

Synonyms: Calcium Trisodium Diethylenetriaminepentaacetate

D1869-25G D1869-5G
TCD1869-025GEA 303.36 CAD
TCD1869-025G TCD1869-5G
Diethylenetriaminepentaacetic acid calcium trisodium salt hydrate ≥98.0%
Diethylenetriaminepentaacetic acid calcium trisodium salt hydrate

CAS Number: 207226-35-5
MDL Number: MFCD00150947
Molecular Formula: C14H23N3O10
Molecular Weight: 497.36
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Other Product Names / Synonyms: Calcium Trisodium Diethylenetriaminepentaacetate

Formula: C₁₄H₂₃N₃O₁₀
MW: 497.36 g/mol
MDL Number: MFCD00150947
CAS Number: 207226-35-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=25aac641cdab9aae1da151ae8eeb934da8b9b95f&vid=25aac641cdab9aae1da151ae8eeb934da8b9b95f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR