(±)-1,2-Dodecanediol ≥93.0%

Supplier: TCI America

D1861-500G D1861-25G
TCD1861-500GEA 554.58 CAD
TCD1861-500G TCD1861-025G
(±)-1,2-Dodecanediol ≥93.0%
(±)-1,2-Dodecanediol

CAS Number: 1119-87-5
MDL Number: MFCD00004726
Molecular Formula: C12H26O2
Molecular Weight: 202.34
Purity/Analysis Method: >93.0% (GC)
Form: Crystal
Melting point (°C): 58


Other Product Names / Synonyms: 1,2-Dihydroxydodecane

Formula: C₁₂H₂₆O₂
Melting Pt: 58 °C
MDL Number: MFCD00004726
CAS Number: 1119-87-5
EINECS: 214-289-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3dd5b9b2ce657c89540a9a13a45e328ea93f9d75&vid=3dd5b9b2ce657c89540a9a13a45e328ea93f9d75&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR