4,5-Dimethyl-3-hydroxy-2(5H)-furanone ∼14% ∼1.2 mol/L in propylene glycol

Supplier: TCI America

Synonyms: 3-Hydroxy-4,5-dimethylfuran-2(5H)-one, 4,5-Dimethyl-3-hydroxy-2,5-dihydrofuran-2-one

D1851-25ML D1851-5ML
TCD1851-025MLEA 290.88 CAD
TCD1851-025ML TCD1851-005ML
4,5-Dimethyl-3-hydroxy-2(5H)-furanone ∼14% ∼1.2 mol/L in propylene glycol
4,5-Dimethyl-3-hydroxy-2(5H)-furanone

CAS Number: 28664-35-9
MDL Number: MFCD00059957
Molecular Formula: C6H8O3
Molecular Weight: 128.13
Form: Clear Liquid
Color: Very Pale Yellow
Flash Point (°C): 104

Formula: C₆H₈O₃
MW: 128.13 g/mol
Flash Pt: 104 °C
MDL Number: MFCD00059957
CAS Number: 28664-35-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=973c2307ff403e430187698275369d36bc00706f&vid=89d0faea43e5c7ef2a0ec5a3f10fe592642b029e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR