4,6-Dimethyl-2-hydroxypyrimidine hydrochloride ≥98.0%

Supplier: TCI America

Synonyms: 2-Hydroxy-4,6-dimethylpyrimidine Hydrochloride, 4,6-Dimethyl-2-pyrimidinol Hydrochloride, 4,6-Dimethylpyrimidin-2-ol hydrochloride

D1823-250G D1823-25G
TCD1823-250GEA 325.48 CAD
TCD1823-250G TCD1823-025G
4,6-Dimethyl-2-hydroxypyrimidine hydrochloride ≥98.0%
4,6-Dimethyl-2-hydroxypyrimidine hydrochloride

CAS Number: 34289-60-6
MDL Number: MFCD00035312
Molecular Formula: C6H8N2O
Molecular Weight: 160.60
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 281


Other Product Names / Synonyms: 4,6-Dimethyl-2-pyrimidinol Hydrochloride, 2-Hydroxy-4,6-dimethylpyrimidine Hydrochloride

Formula: C₆H₈N₂O
MW: 160.60 g/mol
Melting Pt: 281 °C
MDL Number: MFCD00035312
CAS Number: 34289-60-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f0932f95a81ec00a9895ddb12010dd557e0a78e&vid=3f0932f95a81ec00a9895ddb12010dd557e0a78e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR