1,3-Diphenyltetramethyldisilazane ≥95.0%
Supplier: TCI America
D1816-5ML
D1816-25ML
TCD1816-005MLEA
142.2
CAD
TCD1816-005ML
TCD1816-25ML
1,3-Diphenyltetramethyldisilazane ≥95.0%
1,3-Diphenyltetramethyldisilazane
CAS Number: 3449-26-1
MDL Number: MFCD00042916
Molecular Formula: C16H23NSi2
Molecular Weight: 285.54
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 146
Flash Point (°C): 162
Specific Gravity (20/20): 0.99
Product Categories: Analytical Science, Silylation (GC Derivatizing Reagents)
Other Product Names / Synonyms: Tetramethyl-1,3-diphenyldisilazane
Formula:
C₁₆H₂₃NSi₂ Boiling Pt: 146 °C Density: 0.99 (20 °C) Flash Pt: 162 °C |
MDL Number:
MFCD00042916 CAS Number: 3449-26-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...