3,6-Dihydroxybenzonorbornane ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
D1806-25G
D1806-5G
TCD1806-25GEA
737.86
CAD
TCD1806-25G
TCD1806-005G
3,6-Dihydroxybenzonorbornane ≥98.0% (by HPLC, titration analysis)
3,6-Dihydroxybenzonorbornane
[Antioxidant]
CAS Number: 16144-91-5
MDL Number: MFCD00077441
Molecular Formula: C11H12O2
Molecular Weight: 176.22
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 180
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Benzonorbornene-3,6-diol, 1,2,3,4-Tetrahydro-1,4-methanonaphthalene-5,8-diol
Formula:
C₁₁H₁₂O₂ Melting Pt: 180 °C |
MDL Number:
MFCD00077441 CAS Number: 16144-91-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...