1,1-Dimethylguanidinium sulfate ≥98.0% for synthesis

Supplier: TCI America

Synonyms: N,N-Dimethylguanidine sulfate, 1,1-Dimethylguanidine Sulfate(2:1), N,N-Dimethylguanidine sulphate, 1,1-Dimethylguanidine sulphate(2:1)

D1799-25G D1799-5G
TCD1799-025GEA 115.34 CAD
TCD1799-025G 76218-984
1,1-Dimethylguanidinium sulfate ≥98.0% for synthesis
1,1-Dimethylguanidinium sulphate

CAS Number: 598-65-2
MDL Number: MFCD00013131
Molecular Formula: C3H9N3
Molecular Weight: 272.32
Purity/Analysis Method: >98.0% (T)
Form: Crystal

Formula: C₆H₂₀N₆O₄S
MDL Number: MFCD00013131
CAS Number: 598-65-2
EINECS: 209-946-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=02f9c73cca305f90b395bf1cc67197a3e174fcb2&vid=02f9c73cca305f90b395bf1cc67197a3e174fcb2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR