N-[3-(Dimethylamino)propyl]acrylamide 98,0 Stabilised

Supplier: TCI America

D1785-25G D1785-500G D1785-100G
TCD1785-025GEA 58.46 CAD
TCD1785-025G TCD1785-500G TCD1785-100G
N-[3-(Dimethylamino)propyl]acrylamide 98,0 Stabilised
N-[3-(Dimethylamino)propyl]acrylamide

(stabilized with MEHQ)
CAS Number: 3845-76-9
MDL Number: MFCD00059902
Molecular Formula: C8H16N2O
Molecular Weight: 156.23
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 142
Flash Point (°C): 140
Specific Gravity (20/20): 0.95
Storage Temperature: 0-10°C

Formula: C₈H₁₆N₂O
MW: 156.23 g/mol
Boiling Pt: 142 °C
Density: 0.95 (20 °C)
Flash Pt: 140 °C
MDL Number: MFCD00059902
CAS Number: 3845-76-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ef72a0ef7ddd5cf9dbf9d57155a6fec3376b6d67&vid=ef72a0ef7ddd5cf9dbf9d57155a6fec3376b6d67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR