Nickel(II) dibutyldithiocarbamate ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Nickel(II) Bis(dibutyldithiocarbamate), Dibutyldithiocarbamic Acid Nickel(II) Salt

D1781-25G D1781-500G
TCD1781-025GEA 56.88 CAD
TCD1781-025G TCD1781-500G
Nickel(II) dibutyldithiocarbamate ≥97.0% (by titrimetric analysis)
Nickel(II) dibutyldithiocarbamate

CAS Number: 13927-77-0
MDL Number: MFCD00067273
Molecular Formula: C9H19NS2
Molecular Weight: 467.43
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 89


Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: Dibutyldithiocarbamic Acid Nickel(II) Salt, Nickel(II) Bis(dibutyldithiocarbamate)

Formula: C₉H₁₉NS₂
MW: 467.43 g/mol
Melting Pt: 89 °C
MDL Number: MFCD00067273
CAS Number: 13927-77-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6634d9a829c641718706923e490f85160ba8fa27&vid=6634d9a829c641718706923e490f85160ba8fa27&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR