Stearyl acrylate ≥97.0% stabilised

Supplier: TCI America

Synonyms: Octadecyl Acrylate, Acrylic Acid Stearyl Ester, Acrylic Acid Octadecyl Ester

A1011-25G A1011-400G
TCA1011-025GEA 42.66 CAD
TCA1011-025G TCA1011-400G
Stearyl acrylate ≥97.0% stabilised
Stearyl acrylate

(stabilized with MEHQ)
CAS Number: 4813-57-4
MDL Number: MFCD00015091
Molecular Formula: C21H40O2
Molecular Weight: 324.55
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 28
Flash Point (°C): 191


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Acrylic Acid Octadecyl Ester, Acrylic Acid Stearyl Ester, Octadecyl Acrylate

Formula: C₂₁H₄₀O₂
Melting Pt: 28 °C
Flash Pt: 191 °C
MDL Number: MFCD00015091
CAS Number: 4813-57-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=640094ef7740b9e30098908b5c3008a2fb46f510&vid=640094ef7740b9e30098908b5c3008a2fb46f510&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR