4,4'-Dithiodibutyric acid ≥95.0% (by GC, titration analysis)

Supplier: TCI America

D1757-25G D1757-5G
TCD1757-025GEA 267.04 CAD
TCD1757-025G TCD1757-5G
4,4'-Dithiodibutyric acid ≥95.0% (by GC, titration analysis)
4,4'-Dithiodibutyric acid

CAS Number: 2906-60-7
MDL Number: MFCD00004406
Molecular Formula: C8H14O4S2
Molecular Weight: 238.32
Purity/Analysis Method: >95.0% (GC,T)
Form: Crystal
Melting point (°C): 108


Other Product Names / Synonyms: Bis(3-carboxypropyl) Disulfide, 3-Carboxypropyl Disulfide

Formula: C₈H₁₄O₄S₂
Melting Pt: 108 °C
MDL Number: MFCD00004406
CAS Number: 2906-60-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1260bfb11533ad5a37604e44330c509d766fce4e&vid=1260bfb11533ad5a37604e44330c509d766fce4e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR