2-[2-(Dimethylamino)ethoxy]ethanol ≥98.0%

Supplier: TCI America

Synonyms: Ethylene Glycol Mono[2-(dimethylamino)ethyl] Ether, 2-(2-(Dimethylamino)ethoxy)ethanol

D1756-25ML D1756-500ML
TCD1756-025MLEA 28.44 CAD
TCD1756-025ML TCD1756-500ML
2-[2-(Dimethylamino)ethoxy]ethanol ≥98.0%
2-[2-(Dimethylamino)ethoxy]ethanol

CAS Number: 1704-62-7
MDL Number: MFCD00059602
Molecular Formula: C6H15NO2
Molecular Weight: 133.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 95
Flash Point (°C): 93
Specific Gravity (20/20): 0.95


Product Categories: Materials Chemistry, Polymerization Catalysts
Other Product Names / Synonyms: Ethylene Glycol Mono[2-(dimethylamino)ethyl] Ether

Formula: C₆H₁₅NO₂
MW: 133.19 g/mol
Boiling Pt: 95 °C
Density: 0.95 (20 °C)
Flash Pt: 93 °C
MDL Number: MFCD00059602
CAS Number: 1704-62-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ba0d5c19a69b8099ad78b3984dbb845a29d7b08&vid=0ba0d5c19a69b8099ad78b3984dbb845a29d7b08&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR